Merck
  • Latent transforming growth factor-β1 protects against bleomycin-induced lung injury in mice.
Latent transforming growth factor-β1 protects against bleomycin-induced lung injury in mice.
American journal of respiratory cell and molecular biology (2014-06-03)
Yong-Jiang Tang, Jun Xiao, Xiao Ru Huang, Yang Zhang, Chen Yang, Xiao-Ming Meng, Yu-Lin Feng, Xiao-Jing Wang, David S C Hui, Cheuk-Man Yu, Hui Yao Lan
ABSTRACT
Transforming growth factor (TGF)-β1 is a potent mediator known to induce lung fibrosis. However, the role of latent TGF-β1 in lung inflammation and fibrosis is unclear. To investigate the role of circulating latent TGF-β1 in bleomycin-induced lung injury, lung disease was induced in keratin-5 promoter-driven TGF-β1(wt) transgenic (Tg) mice by bleomycin. The role of latent TGF-β1 in pulmonary inflammation and fibrosis was examined at Days 7 and 28 after administration of bleomycin. Compared with littermate wild-type (WT) mice, TGF-β1(wt) Tg mice had over twofold-higher levels of latent TGF-β1 in both plasma and lung tissue, and were protected from bleomycin-induced pulmonary inflammation, such as up-regulation of IL-1β, TNF-α, and macrophage chemotactic protein-1, and infiltration of CD3(+) T cells and F4/80(+) macrophages. In addition, the severity of lung fibrosis with massive collagen matrix accumulation was markedly reduced in TGF-β1(wt) Tg mice. These protective effects were associated with higher levels of Smad7 and inactivation of both NF-κB and TGF-β/Smad3 signaling pathways, in addition to an increase in forkhead box P3 (Foxp3)-dependent regulatory T cells, but inhibition of T helper 17-mediated lung injury. In summary, mice overexpressing latent TGF-β1 are protected from bleomycin-induced lung injury. Triggering the Smad7 negative feedback mechanism to inhibit both NF-κB and TGF-β/Smad signaling pathways, and enhancing the regulatory T cell response to counter-regulate T helper 17-mediated lung injury, are potential mechanisms by which latent TGF-β1 protects against bleomycin-induced lung injury.
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, suitable for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Coronatine, from Pseudomonas syringae pv Glycinea, >95% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Bleomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%