Merck
  • Unraveling the contribution of ectoenzymes to myeloma life and survival in the bone marrow niche.
Unraveling the contribution of ectoenzymes to myeloma life and survival in the bone marrow niche.
Annals of the New York Academy of Sciences (2014-07-23)
Valeria Quarona, Valentina Ferri, Antonella Chillemi, Marina Bolzoni, Cristina Mancini, Gianluca Zaccarello, Ilaria Roato, Fabio Morandi, Danilo Marimpietri, Giuliano Faccani, Eugenia Martella, Vito Pistoia, Nicola Giuliani, Alberto L Horenstein, Fabio Malavasi
ABSTRACT
The bone marrow provides a protected environment for generating a vast array of cell types. Bones are thus a dynamic source of structural components and soluble factors used either locally or at a distance from their site of production. We discuss the role of ectoenzymes in the bone niche where human myeloma grows. Selected ectoenzymes have been tested for their ability to promote production of substrates involved in signaling, synthesis of growth factors and hormones, and modulation of the immune response. Because of the difficulty of simultaneously tracking all these activities, we narrow our focus to events potentially influencing synthesis of adenosine (ADO), an important regulator of multiple biological functions, including local immunological tolerance. Our working hypothesis, to be discussed and partially tested herein, is that CD38, and likely BST1/CD157--both NAD(+) -consuming enzymes, are active in the myeloma niche and lead a discontinuous chain of ectoenzymes whose final products are exploited by the neoplastic plasma cell as part of its local survival strategy. Coadjuvant ectoenzymes include PC-1/CD203a, CD39, and CD73, which control the production of ADO. Results discussed here and from ongoing experiments indicate that the myeloma niche hosts the canonical, as well as alternative, pathways of ADO generation. Other possibilities are presented and discussed.
Product Description
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
NADase from porcine brain, ≥0.007 unit/mg solid
Sigma-Aldrich
Phosphodiesterase I from Crotalus adamanteus venom, Type VI, crude dried venom
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine