Merck
  • Drug-induced changes to the vertebral endplate vasculature affect transport into the intervertebral disc in vivo.
Drug-induced changes to the vertebral endplate vasculature affect transport into the intervertebral disc in vivo.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2014-09-05)
Sarah E Gullbrand, Joshua Peterson, Rosemarie Mastropolo, James P Lawrence, Luciana Lopes, Jeffrey Lotz, Eric H Ledet
ABSTRACT
Intervertebral disc health is mediated in part by nutrient diffusion from the microvasculature in the adjacent subchondral bone. Evidence suggests that a reduction in nutrient diffusion contributes to disc degeneration, but the role of the microvasculature is unclear. The purpose of this study was to induce changes in the endplate microvasculature in vivo via pharmaceutical intervention and then correlate microvasculature characteristics to diffusion and disc health. New Zealand white rabbits were administered either nimodipine (to enhance microvessel density) or nicotine (to diminish microvessel density) daily for 8 weeks compared to controls. Trans-endplate diffusion and disc health were quantified using post-contrast enhanced magnetic resonance imaging (MRI). Histology was utilized to assess changes to the subchondral vasculature. Results indicate that nimodipine increased vessel area and vessel-endplate contact length, causing a significant increase in disc diffusion. Surprisingly, nicotine caused increases in vessel number and area but did not alter diffusion into the disc. The drug treatments did affect the microvasculature and diffusion, but the relationship between the two is complex and dependent on multiple factors which include vessel-endplate distance, and vessel-endplate contact length in addition to vessel density. Our data suggest that drugs can modulate these factors to augment or diminish small molecule transport.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Nimodipine
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Nimodipine, Pharmaceutical Secondary Standard; Certified Reference Material
Nimodipine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology