Merck
  • Advanced stage anaplastic large cell lymphoma in children and adolescents: results of ANHL0131, a randomized phase III trial of APO versus a modified regimen with vinblastine: a report from the children's oncology group.
Advanced stage anaplastic large cell lymphoma in children and adolescents: results of ANHL0131, a randomized phase III trial of APO versus a modified regimen with vinblastine: a report from the children's oncology group.
Pediatric blood & cancer (2014-08-27)
Sarah Alexander, Jacqueline M Kraveka, Sheila Weitzman, Eric Lowe, Lynette Smith, James C Lynch, Myron Chang, Marsha C Kinney, Sherrie L Perkins, Joseph Laver, Thomas G Gross, Howard Weinstein
ABSTRACT
Optimal therapy for children and adolescents with advanced stage anaplastic large cell lymphoma (ALCL) is unknown. ANHL0131 examined whether a maintenance regimen including vinblastine compared to the standard APO (doxorubicin, prednisone, vincristine, methotrexate, 6-mercaptopurine) regimen would result in superior event-free survival. One hundred and twenty five eligible patients were enrolled. Induction was identical for both arms. Post induction patients were randomized to receive APO with vincristine every 3 weeks or a regimen that substituted vincristine with weekly vinblastine (APV). There was no difference between the patients randomized to the APO versus APV arms in either event free survival (EFS) or overall survival (OS) (three year EFS 74% vs. 79%, P = 0.68 and three years OS of 84% vs. 86%, P = 0.87, respectively). Patients in the APV arm required dose reduction secondary to myelosuppression and had a higher incidence of neutropenia as well as infection with neutropenia compared to those in the APO arm (P < 0.001, P = 0.019, respectively). Treatment with weekly vinblastine instead of every three week vincristine as part of multi-agent maintenance therapy did not result in improvement in EFS or OS. Weekly vinblastine was associated with increased toxicity. (ClinicalTrials.gov Identifier NCT00059839).
Product Description
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Methotrexate
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
6-Mercaptopurine monohydrate, 98%
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard