Merck
  • Multicenter study of intravitreal dexamethasone implant in noninfectious uveitis: indications, outcomes, and reinjection frequency.
Multicenter study of intravitreal dexamethasone implant in noninfectious uveitis: indications, outcomes, and reinjection frequency.
American journal of ophthalmology (2014-09-15)
Javier Zarranz-Ventura, Ester Carreño, Robert L Johnston, Quresh Mohammed, Adam H Ross, Carl Barker, Alex Fonollosa, Joseba Artaraz, Laura Pelegrin, Alfredo Adan, Richard W Lee, Andrew D Dick, Ahmed Sallam
ABSTRACT
To identify clinical outcomes and treatment patterns of intravitreal dexamethasone implant (Ozurdex; Allergan, Inc) in noninfectious uveitis in the clinical setting. Multicenter retrospective cohort study. Eighty-two eyes (63 patients) receiving 142 implant injections over 35 months were included. Treatment indication, uveitis diagnosis, visual acuity, intraocular pressure, vitreous haze score, central retinal thickness by optical coherence tomography, phakic status, number of injections, time to reinjection, systemic treatments, and complications data were collected. Time to visual acuity and vitreous haze score improvement as per the Standardization of Uveitis Nomenclature guidelines were also determined. The probability of visual acuity improvement (≥0.3 logarithm of the minimal angle of resolution units improvement) was 39% at 1 month, 49% at 3 months, 52% at 6 months, and 58% at 12 months. Eyes with baseline vitritis (vitreous haze score ≥+0.5, n = 45) had a probability of vitreous haze score improvement (2-step decrease or change from +0.5 to 0) at 2 weeks of 41%, at 1 month 63%, at 3 months 73%, at 6 months 79%, and at 12 months 88%. In eyes that completed 12-month follow-up (n = 54), 40.7% underwent 2 injections (mean time to second injection of 6.6 ± 1.9 months) and 11.2% required ≥3 injections (mean time to third injection of 11 ± 1.5 months). Dexamethasone implant use in uveitis provides favorable visual acuity and vitreous haze score outcomes but requires repeated injections, an important consideration when choosing intraocular treatment as a route to controlling uveitis.
Product Description
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard