Merck
  • Lactate dehydrogenase is the key enzyme for pneumococcal pyruvate metabolism and pneumococcal survival in blood.
Lactate dehydrogenase is the key enzyme for pneumococcal pyruvate metabolism and pneumococcal survival in blood.
Infection and immunity (2014-09-24)
Paula Gaspar, Firas A Y Al-Bayati, Peter W Andrew, Ana Rute Neves, Hasan Yesilkaya
ABSTRACT
Streptococcus pneumoniae is a fermentative microorganism and causes serious diseases in humans, including otitis media, bacteremia, meningitis, and pneumonia. However, the mechanisms enabling pneumococcal survival in the host and causing disease in different tissues are incompletely understood. The available evidence indicates a strong link between the central metabolism and pneumococcal virulence. To further our knowledge on pneumococcal virulence, we investigated the role of lactate dehydrogenase (LDH), which converts pyruvate to lactate and is an essential enzyme for redox balance, in the pneumococcal central metabolism and virulence using an isogenic ldh mutant. Loss of LDH led to a dramatic reduction of the growth rate, pinpointing the key role of this enzyme in fermentative metabolism. The pattern of end products was altered, and lactate production was totally blocked. The fermentation profile was confirmed by in vivo nuclear magnetic resonance (NMR) measurements of glucose metabolism in nongrowing cell suspensions of the ldh mutant. In this strain, a bottleneck in the fermentative steps is evident from the accumulation of pyruvate, revealing LDH as the most efficient enzyme in pyruvate conversion. An increase in ethanol production was also observed, indicating that in the absence of LDH the redox balance is maintained through alcohol dehydrogenase activity. We also found that the absence of LDH renders the pneumococci avirulent after intravenous infection and leads to a significant reduction in virulence in a model of pneumonia that develops after intranasal infection, likely due to a decrease in energy generation and virulence gene expression.
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Lactic acid, 88%, FCC
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Galactose, European Pharmacopoeia (EP) Reference Standard
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
USP
Lactic acid, United States Pharmacopeia (USP) Reference Standard
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Pyruvic acid, natural, ≥95%, FG
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Sigma-Aldrich
Pyruvic acid, 95%
Sigma-Aldrich
Pyruvic acid, 98%
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup