Merck
  • Construction of monomer-free, highly crosslinked, water-compatible polymers.
Construction of monomer-free, highly crosslinked, water-compatible polymers.
Journal of dental research (2014-09-25)
E A Dailing, S H Lewis, M D Barros, J W Stansbury
ABSTRACT
Polymeric dental adhesives require the formation of densely crosslinked network structures to best ensure mechanical strength and durability in clinical service. Monomeric precursors to these materials typically consist of mixtures of hydrophilic and hydrophobic components that potentially undergo phase separation in the presence of low concentrations of water, which is detrimental to material performance and has motivated significant investigation into formulations that reduce this effect. We have investigated an approach to network formation based on nanogels that are dispersed in inert solvent and directly polymerized into crosslinked polymers. Monomers of various hydrophilic or hydrophobic characteristics were copolymerized into particulate nanogels bearing internal and external polymerizable functionality. Nanogel dispersions were stable at high concentrations in acetone or, with some exceptions, in water and produced networks with a wide range of mechanical properties. Networks formed rapidly upon light activation and reached high conversion with extremely low volumetric shrinkage. Prepolymerizing monomers into reactive nanostructures significantly changes how hydrophobic materials respond to water compared with networks obtained from polymerizations involving free monomer. The modulus of fully hydrated networks formed solely from nanogels was shown to equal or exceed the modulus in the dry state for networks based on nanogels containing a hydrophobic dimethacrylate and hydrophilic monomethacrylate, a result that was not observed in a hydroxyethyl methacrylate (HEMA) homopolymer or in networks formed from nanogels copolymerized with HEMA. These results highlight the unique approach to network development from nanoscale precursors and properties that have direct implications in functional dental materials.
Product Description
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Hydroxyethyl methacrylate
Supelco
Acetone, analytical standard
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), purum, ≥98.0% (GC)
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
2-Isocyanatoethyl methacrylate, contains ≤0.1% BHT as inhibitor, 98%
Supelco
2-Butanone, analytical standard
Sigma-Aldrich
Tetraethylene glycol dimethacrylate, technical, ≥90% (GC)
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%