Merck
  • EGFR activation and signaling in cancer cells are enhanced by the membrane-bound metalloprotease MT4-MMP.
EGFR activation and signaling in cancer cells are enhanced by the membrane-bound metalloprotease MT4-MMP.
Cancer research (2014-10-17)
Alexandra Paye, Alice Truong, Cassandre Yip, Jonathan Cimino, Silvia Blacher, Carine Munaut, Didier Cataldo, Jean Michel Foidart, Erik Maquoi, Joelle Collignon, Philippe Delvenne, Guy Jerusalem, Agnès Noel, Nor Eddine Sounni
ABSTRACT
MT4-MMP (MMP-17) is a glycosylphosphatidyl inositol-anchored matrix metalloprotease expressed on the surface of cancer cells that promotes tumor growth and metastasis. In this report, we identify MT4-MMP as an important driver of cancer cell proliferation through CDK4 activation and retinoblastoma protein inactivation. We also determine a functional link between MT4-MMP and the growth factor receptor EGFR. Mechanistic experiments revealed direct association of MT4-MMP and its positive effects on EGFR phosphorylation in response to TGFα and EGF in cancer cells. Notably, the effects of MT4-MMP on proliferation and EGFR activation did not rely on metalloprotease activity. Clinically, MT4-MMP and EGFR expressions were correlated in human triple-negative breast cancer specimens. Altogether, our results identify MT4-MMP as a positive modifier of EGFR outside-in signaling that acts to cooperatively drive cancer cell proliferation.
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material