Merck
  • Zinc transporter-1: a novel NMDA receptor-binding protein at the postsynaptic density.
Zinc transporter-1: a novel NMDA receptor-binding protein at the postsynaptic density.
Journal of neurochemistry (2014-10-17)
Manuela Mellone, Silvia Pelucchi, Lorenzo Alberti, Armando A Genazzani, Monica Di Luca, Fabrizio Gardoni
ABSTRACT
Zinc (Zn(2+) ) is believed to play a relevant role in the physiology and pathophysiology of the brain. Hence, Zn(2+) homeostasis is critical and involves different classes of molecules, including Zn(2+) transporters. The ubiquitous Zn(2+) transporter-1 (ZNT-1) is a transmembrane protein that pumps cytosolic Zn(2+) to the extracellular space, but its function in the central nervous system is not fully understood. Here, we show that ZNT-1 interacts with GluN2A-containing NMDA receptors, suggesting a role for this transporter at the excitatory glutamatergic synapse. First, we found that ZNT-1 is highly expressed at the hippocampal postsynaptic density (PSD) where NMDA receptors are enriched. Two-hybrid screening, coimmunoprecipitation experiments and clustering assay in COS-7 cells demonstrated that ZNT-1 specifically binds the GluN2A subunit of the NMDA receptor. GluN2A deletion mutants and pull-down assays indicated GluN2A(1390-1464) domain as necessary for the binding to ZNT-1. Most importantly, ZNT-1/GluN2A complex was proved to be dynamic, since it was regulated by induction of synaptic plasticity. Finally, modulation of ZNT-1 expression in hippocampal neurons determined a significant change in dendritic spine morphology, PSD-95 clusters and GluN2A surface levels, supporting the involvement of ZNT-1 in the dynamics of excitatory PSD. Zn(2+) transporter-1 (ZNT-1) pumps cytosolic Zn(2+) to the extracellular space, but its function in the central nervous system is not fully understood. We show that ZNT-1 interacts with GluN2A-containing NMDA receptors at the glutamatergic synapse. Most importantly, ZNT-1/GluN2A complex is regulated by induction of synaptic plasticity. Modulation of ZNT-1 expression in hippocampal neurons determined a shrinkage of dendritic spines and a reduction of GluN2A surface levels supporting the involvement of ZNT-1 in the dynamics of the excitatory synapse.
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Zinc, wire reel, 5m, diameter 1.0mm, 99.99+%
Zinc, wire reel, 1m, diameter 0.025mm, as drawn, 99%
Zinc, wire reel, 20m, diameter 3.0mm, extruded, 99.9%
Zinc, wire reel, 0.2m, diameter 6.35mm, 99.99+%
Zinc, wire reel, 2m, diameter 2.0mm, 99.99+%
Zinc, wire reel, 1m, diameter 0.05mm, as drawn, 99%
Zinc, wire reel, 25m, diameter 0.25mm, as drawn, 99.99+%
Zinc, wire reel, 0.5m, diameter 1.0mm, 99.99+%
Zinc, wire reel, 25m, diameter 0.05mm, as drawn, 99%
Zinc, wire reel, 2m, diameter 0.025mm, as drawn, 99%
Zinc, wire reel, 5m, diameter 0.125mm, as drawn, 99%
Zinc, wire reel, 10m, diameter 0.125mm, as drawn, 99%
Zinc, wire reel, 1m, diameter 0.5mm, as drawn, 99.99+%
Zinc, wire reel, 0.2m, diameter 1.0mm, 99.999%
Zinc, wire reel, 500m, diameter 1.0mm, extruded, 99.9%
Zinc, wire reel, 1m, diameter 1.0mm, 99.999%
Zinc, wire reel, 50m, diameter 0.25mm, as drawn, 99.99+%
Zinc, wire reel, 1m, diameter 2.0mm, 99.99+%
Zinc, wire reel, 5m, diameter 2.0mm, 99.99+%
Zinc, wire reel, 0.5m, diameter 2.0mm, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.5mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.95+%