Merck
The impact of plant volatiles on bacterial quorum sensing.
Letters in applied microbiology (2014-10-28)
A Ahmad, A M Viljoen, H Y Chenia
ABSTRACT
Studies describing the use of essential oil constituents as antimicrobial agents have steadily increased; however, some phyto-constituents are often overlooked due to unfavourable minimum inhibitory concentration (MIC) values. Virulence depends on transcriptional factors which are regulated by cell-to-cell communication called quorum sensing (QS). This study was undertaken to evaluate the antimicrobial and anti-QS properties of 29 compounds commonly found in essential oils using two bioreporter strains. QS-inhibitory activity was assessed qualitatively by agar diffusion and quantitatively by spectrophotometric assays. MICs of all the tested compounds ranged from 0·032 to >5 mg ml(-1). Twenty-two compounds displayed varying levels of QS inhibitory activity with zones of violacein inhibition ranging from 9 to 16 mm. Majority of tested molecules inhibited violacein and pyocyanin production in Chromobacterium violaceum and Pseudomonas aeruginosa, while seven compounds increased violacein and pyocyanin production. Interestingly, it was observed that the (+)-enantiomers of carvone, limonene and borneol increased violacein and pyocyanin production, while their levorotary analogues inhibited this production. α-Terpineol and cis-3-nonen-1-ol exhibited >90% violacein inhibition, suggesting their potential as QS inhibitors. This preliminary study indicates that plant volatiles have the potential to impede or promote bacterial communication and further studies need to be undertaken to explore the contribution of structural analogues and stereochemistry of molecules in this process. Antimicrobial and antiquorum sensing (QS) properties of 29 common essential oil compounds were evaluated. Interruption of QS may lead to the development of therapeutic, antivirulence agents to control disease-causing pathogens which are preferable over antimicrobial agents as the latter drives selection pressure on microbial communities to acquire resistance. Twenty-two compounds inhibited QS, while seven promoted the QS to a variable extent in Chromobacterium violaceum and Pseudomonas aeruginosa. Preliminary results suggest that QS-inhibitory compounds of natural origin may inspire the formulation of new generation of antimicrobial agents to control infectious pathogens.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyocyanin, Ready Made Solution from Pseudomonas aeruginosa, 5 mg/mL in DMSO
Sigma-Aldrich
Pyocyanin, from Pseudomonas aeruginosa, ≥98% (HPLC)
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Violacein from Janthinobacterium lividum, >98% (violacein (minimum 85% violacein) and deoxyviolacein, HPLC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%