Merck
  • Trivalent chromium inhibits TSP-1 expression, proliferation, and O-GlcNAc signaling in vascular smooth muscle cells in response to high glucose in vitro.
Trivalent chromium inhibits TSP-1 expression, proliferation, and O-GlcNAc signaling in vascular smooth muscle cells in response to high glucose in vitro.
American journal of physiology. Cell physiology (2014-10-31)
Rituparna Ganguly, Soumyadip Sahu, Ronaldo J Chavez, Priya Raman
ABSTRACT
Trivalent chromium (Cr(3+)) is a mineral nutrient reported to have beneficial effects in glycemic and cardiovascular health. In vitro and in vivo studies suggest that Cr(3+) supplementation reduces the atherogenic potential and lowers the risk of vascular inflammation in diabetes. However, effects of Cr(3+) in vascular cells under conditions of hyperglycemia, characteristic of diabetes, remain unknown. In the present study we show that a therapeutically relevant concentration of Cr(3+) (100 nM) significantly downregulates a potent proatherogenic matricellular protein, thrombospondin-1 (TSP-1), in human aortic smooth muscle cells (HASMC) stimulated with high glucose in vitro. Promoter-reporter assays reveal that this downregulation of TSP-1 expression by Cr(3+) occurs at the level of transcription. The inhibitory effects of Cr(3+) on TSP-1 were accompanied by significant reductions in O-glycosylation of cytoplasmic and nuclear proteins. Using Western blotting and immunofluorescence studies, we demonstrate that reduced protein O-glycosylation by Cr(3+) is mediated via inhibition of glutamine: fructose 6-phosphate amidotransferase, a rate-limiting enzyme of the hexosamine pathway, and O-linked N-acetylglucosamine (O-GlcNAc) transferase, a distal enzyme in the pathway that controls intracellular protein O-glycosylation. Additionally, we found that Cr(3+) attenuates reactive oxygen species formation in glucose-stimulated HASMC, suggesting an antioxidant effect. Finally, we report an antiproliferative effect of Cr(3+) that is specific for high glucose and conditions triggering elevated protein O-glycosylation. Taken together, these findings provide the first cellular evidence for a novel role of Cr(3+) to modulate aberrant vascular smooth muscle cell function associated with hyperglycemia-induced vascular complications.
Product Description
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Chromium, foil, light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Chromium, rod, 100mm, diameter 2.0mm, 99.7+%
Chromium, rod, 100mm, diameter 5.0mm, 99.7+%
Chromium, foil, not light tested, 50x50mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Chromium, rod, 10mm, diameter 2.0mm, 99.7+%
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Chromium, bar, 100mm x 2mm x 2mm, 99.7+%
Chromium, foil, 100x100mm, thickness 3.0mm, hard, 99.95%
Chromium, foil, 25x25mm, thickness 2.0mm, hard, 99.7+%
Chromium, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chromium, foil, 50x50mm, thickness 2.0mm, hard, 99.7+%
Chromium, microfoil, disks, 10mm, thinness 0.025μm, specific density 18.21μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, microfoil, disks, 10mm, thinness 0.25μm, specific density 178.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Chromium, rod, 10mm, diameter 5.0mm, 99.7+%
Chromium, microfoil, disks, 10mm, thinness 0.05μm, specific density 37.27μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.99+%