Merck
  • Peppermint (Mentha piperita) and albendazole against anisakiasis in an animal model.
Peppermint (Mentha piperita) and albendazole against anisakiasis in an animal model.
Tropical medicine & international health : TM & IH (2014-10-21)
Ma Carmen Romero, Ma Concepción Navarro, Joaquina Martín-Sánchez, Adela Valero
ABSTRACT
Therapy against anisakiasis requires invasive techniques to extract L3 , and an effective drug against this nematode is needed. The aim of this study was to determine the efficacy of peppermint essential oil (EO) and its main components against the parasite in comparison to albendazole, a drug currently prescribed to treat anisakiasis. We conducted in vitro experiments and studied an experimental model simulating the human infection in Wistar rats. We used polymerase chain reaction restriction fragment length polymorphism to identify A. simplex s.s. and A. pegreffii and determine any differences in their pathogenicity and susceptibility to the treatments. The in vitro and in vivo experiments both showed that the larvicidal activity of peppermint EO, menthol, menthone and menthyl acetate is higher than that of albendazole. Large stomach lesions were observed in 46.7% of the albendazole-treated rats, whereas no gastrointestinal lesions were detected in those treated with peppermint EO, menthol, menthyl acetate or menthone. In this animal model, treatment with peppermint EO or its main components was more effective than was treatment with albendazole. Lesions were more frequently produced by A. simplex s.s. larvae than by A. pegreffii larvae.
Sigma-Aldrich
DL-Menthol, ≥95%, FCC, FG
Sigma-Aldrich
(±)-Menthol, racemic, ≥98.0% (GC)
Supelco
Albendazole, analytical standard, ≥98%
Albendazole, European Pharmacopoeia (EP) Reference Standard
Menthol, European Pharmacopoeia (EP) Reference Standard
Supelco
DL-Menthol, analytical standard
Supelco
Albendazole, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Albendazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Peppermint oil, terpeneless, FCC
Sigma-Aldrich
Peppermint oil, natural, from Mentha piperita L.
Sigma-Aldrich
Menthol, 99%
Sigma-Aldrich
Peppermint oil, purified by triple-distillation, FCC, FG
Sigma-Aldrich
Peppermint oil, natural, FCC, FG