Merck
  • Accidental subretinal brilliant blue G migration during internal limiting membrane peeling surgery.
Accidental subretinal brilliant blue G migration during internal limiting membrane peeling surgery.
JAMA ophthalmology (2014-10-17)
Felipe P P Almeida, Ana Claudia De Lucca, Ingrid Ursula Scott, Rodrigo Jorge, Andre Messias
ABSTRACT
This case report describes a man who developed retinal changes in his right eye associated with brilliant blue G migration into the subretinal space during 2 years of follow-up. The patient's best-corrected visual acuity in the right eye was 20/70 before surgery, and it improved to 20/25 at 1 year after surgery. Fluorescein angiography showed staining during the late phase in the central macula at all follow-up visits after surgery. Multifocal electroretinography demonstrated normal amplitude and implicit times before surgery but decreased amplitudes and increased implicit times in at least 5 contiguous hexagons after surgery on all 3 examinations performed during the 2-year follow-up period. These functional changes were not topographically correlated with the area of fluorescein staining or with the internal limiting membrane peeled area, but were matched to the area where brilliant blue G accidentally entered the subretinal space. Microperimetry demonstrated reduced retinal threshold sensitivity, particularly in areas with decreased multifocal electroretinography amplitude. Despite the visual acuity improvement observed in this case, multifocal electroretinography and microperimetry indicate that subretinal brilliant blue G might cause focal macular damage with a decrease of macular function suggestive of a toxic effect.
Sigma-Aldrich
Brilliant Blue G, 250, suitable for microscopy
Sigma-Aldrich
Brilliant Blue G, pure
Sigma-Aldrich
Brilliant Blue G solution, Concentrate