Merck
  • Improved outcome of adult Burkitt lymphoma/leukemia with rituximab and chemotherapy: report of a large prospective multicenter trial.
Improved outcome of adult Burkitt lymphoma/leukemia with rituximab and chemotherapy: report of a large prospective multicenter trial.
Blood (2014-11-02)
Dieter Hoelzer, Jan Walewski, Hartmut Döhner, Andreas Viardot, Wolfgang Hiddemann, Karsten Spiekermann, Hubert Serve, Ulrich Dührsen, Andreas Hüttmann, Eckhard Thiel, Jolanta Dengler, Michael Kneba, Markus Schaich, Ingo G H Schmidt-Wolf, Joachim Beck, Bernd Hertenstein, Albrecht Reichle, Katarzyna Domanska-Czyz, Rainer Fietkau, Heinz-August Horst, Harald Rieder, Stefan Schwartz, Thomas Burmeister, Nicola Gökbuget
ABSTRACT
This largest prospective multicenter trial for adult patients with Burkitt lymphoma/leukemia aimed to prove the efficacy and feasibility of short-intensive chemotherapy combined with the anti-CD20 antibody rituximab. From 2002 to 2011, 363 patients 16 to 85 years old were recruited in 98 centers. Treatment consisted of 6 5-day chemotherapy cycles with high-dose methotrexate, high-dose cytosine arabinoside, cyclophosphamide, etoposide, ifosphamide, corticosteroids, and triple intrathecal therapy. Patients >55 years old received a reduced regimen. Rituximab was given before each cycle and twice as maintenance, for a total of 8 doses. The rate of complete remission was 88% (319/363); overall survival (OS) at 5 years, 80%; and progression-free survival, 71%; with significant difference between adolescents, adults, and elderly patients (OS rate of 90%, 84%, and 62%, respectively). Full treatment could be applied in 86% of the patients. The most important prognostic factors were International Prognostic Index (IPI) score (0-2 vs 3-5; P = .0005), age-adjusted IPI score (0-1 vs 2-3; P = .0001), and gender (male vs female; P = .004). The high cure rate in this prospective trial with a substantial number of participating hospitals demonstrates the efficacy and feasibility of chemoimmunotherapy, even in elderly patients. This trial was registered at www.clinicaltrials.gov as #NCT00199082.
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
SAFC
Methotrexate
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Ifosfamide, ≥98%
Ifosfamide, European Pharmacopoeia (EP) Reference Standard
Etoposide, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, ≥90% (HPLC), crystalline
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard