Merck
  • Exendin-4 promotes endothelial barrier enhancement via PKA- and Epac1-dependent Rac1 activation.
Exendin-4 promotes endothelial barrier enhancement via PKA- and Epac1-dependent Rac1 activation.
American journal of physiology. Cell physiology (2014-11-08)
Ai Q Li, Liang Zhao, Teng F Zhou, Meng Q Zhang, Xiao M Qin
ABSTRACT
Among emerging antidiabetic agents, glucagon-like peptide-1 (GLP-1)-based therapies carry special cardiovascular implications, exerting both direct and indirect effects. The control of vascular permeability is of pivotal importance in vascular pathologies. The objective of the present study was to determine the effect of GLP-1 on endothelial barrier function and assess the underlying mechanism(s). Here we show for the first time that the stable GLP-1 analog exendin-4 attenuated the leakage of subcutaneous blood vessels in mice indexed by dye extravasation caused by injections of thrombin. Moreover, in cultured endothelial cells, exendin-4 significantly prevented the thrombin-induced FITC-dextran permeability of endothelial monolayers via GLP-1 receptor. Immunofluorescence microscopy reveals that exendin-4 abrogates detrimental effects of thrombin on VE-cadherin and the F-actin cytoskeleton, with decreased stress fiber and gap formation. Importantly, exendin-4 reduced thrombin-induced tyrosine phosphorylation of VE-cadherin at Y731 and Y658. Moreover, small GTPase Rac1 was significantly activated as a result of exendin-4 treatment. The efficacy of exendin-4 to counteract the barrier-compromising effect of thrombin was blunted when Rac1 was inactivated by Rac1 inhibitor NSC-23766. Inhibition of PKA activity or small-interfering RNA for exchange protein directly activated by cAMP 1 (Epac1) decreased exendin-4-induced Rac1 activation and barrier enhancement, indicating the participation of both PKA and Epac1 in the barrier-stabilizing effect of exendin-4 elicited on thrombin-impaired barrier function. Thus, our findings have uncovered an unpredicted role for exendin-4 in the coordination of vascular permeability and clarified the molecular underpinnings that contribute to barrier restoration initiated by exendin-4.
Product Description
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 25,000
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
Dextran, analytical standard, suitable for gel permeation chromatography (GPC), Set Mp 1,000-400,000
Dextran 4 for calibration, Dextran 4 European Pharmacopoeia (EP) Reference Standard
Dextran Vo, Dextran Vo European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Supelco
Dextran, analytical standard, 670,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran, enzymatic synth.
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 50,000
Supelco
Dextran, analytical standard, 25,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Supelco
Dextran, analytical standard, 12,000
Supelco
Dextran, analytical standard, 410,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 5,000
Supelco
Dextran, analytical standard, 1,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)