Merck
  • Acute cadmium administration to rats exerts both immunosuppressive and proinflammatory effects in spleen.
Acute cadmium administration to rats exerts both immunosuppressive and proinflammatory effects in spleen.
Toxicology (2014-12-03)
Jelena Demenesku, Ivana Mirkov, Marina Ninkov, Aleksandra Popov Aleksandrov, Lidija Zolotarevski, Dragan Kataranovski, Milena Kataranovski
ABSTRACT
Conflicting data (both suppression and augmentation as well as lack of the effect) exist in respect to cadmium (Cd) and splenic T cell-based immune cell activity. Spleen is also the site of innate immune responses but impact of Cd on this type of immunity has been less explored. In the present study the effects of acute Cd administration on basic aspects of both T cell-based and innate immune spleen cell activity were examined in rats. Intraperitoneal injection of 1mg of Cd/kg resulted in decrease in concanavalin A (ConA) induced proliferation which seems to be more related to altered spleen cells responsiveness to IL-2 than to apoptosis. Differential effects on proinflammatory T cell derived cytokines were observed (decreases of IFN-γ gene expression and ConA-stimulated production, but increases in IL-17 mRNA levels with no effect on concentrations of protein product). Reduction of IFN-γ production seemed not to rely on IL-4 and IL-10, but at least partly on nitric oxide (NO). Increased activity relevant for innate immunity (granulocyte and CD11b(+) cell accumulation in the spleen, inducible nitric oxide synthase/iNOS expression and NO production by spleen cells) was observed, but there was a decrease in respiratory burst (dihydrorhodamine/DHR oxidation and nitroblue tetrazolium/NBT reduction). Increases of TNF-α and IL-1β gene expression and IL-1β protein product were noted as well. Administration of 0.5mg Cd/kg resulted in less pronounced (ConA-induced proliferation) or lack of the effect (IFN-γ production) on spleen T cell activities and on innate activities (granulocyte accumulation, NO production) as well. However, increases of spleen cell respiratory burst activity and IL-1β production were observed. Effects of lower cadmium doses (5ppm and 50ppm) on several aspects of spleen cell immune activity were observed in intermediate period of exposure (30 days, oral intake) as well. Differential effects of Cd on immune activities of spleen cells might contribute to our understanding of the complexity of immunomodulatory effects of this metal.
Product Description
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sigma-Aldrich
Ethylenediamine dihydrochloride, 98%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
USP
Sulfanilamide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sulfanilamide, ≥98%
USP
Sulfanilamide, United States Pharmacopeia (USP) Reference Standard
Sulfanilamide, European Pharmacopoeia (EP) Reference Standard
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Supelco
Sodium nitrite, analytical standard
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Cadmium chloride
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%