Merck
  • Micro-RNA 21Targets dual specific phosphatase 8 to promote collagen synthesis in high glucose-treated primary cardiac fibroblasts.
Micro-RNA 21Targets dual specific phosphatase 8 to promote collagen synthesis in high glucose-treated primary cardiac fibroblasts.
The Canadian journal of cardiology (2014-11-25)
Shulei Liu, Wenqi Li, Mingtong Xu, Hui Huang, Jingfeng Wang, Xiaochao Chen
ABSTRACT
Micro-RNA 21 (miR-21) has been shown to contribute to cardiac fibrosis in many diseases. In this study we investigated the role of miR-21 in excessive production of collagen in diabetic cardiomyopathy. The proliferation rate of cardiac fibroblasts was analyzed by Western blot, Cell Counting Kit-8 kit (Dojindo Molecular Technologies, Kumamoto, Japan), and Cell-Light EdU Apollo 488 In Vitro Imaging Kit (RiboBio, Guangzhou, China). Real-time polymerase chain reaction and Western blotting were conducted to determine gene expression levels. A luciferase reporter assay was used to verify the interaction between miR-21 and the 3' untranslated region (3'UTR) of dual specific phosphatase 8 (DUSP8). Our results show that high glucose promoted the proliferation and collagen synthesis of rat cardiac fibroblasts, which was accompanied by an increase of miR-21. Gain-of-function and loss-of-function assays confirmed that miR-21 mediated this effect, suggesting the crucial role of miR-21 in diabetic cardiomyopathy. Our study also identified a direct target of miR-21, DUSP8, which regulates cell proliferation and collagen synthesis in cardiac fibroblasts through p38 and c-Jun N-terminal kinase (JNK)/stress-activated kinase (SAPK) signalling. Our results show that miR-21 bound to the 3'UTR of DUSP8 post-transcriptionally repressed its expression. In addition, enforced expression of miR-21 activated the JNK/SAPK and p38 signalling pathways. Our study shows that miR-21 promotes high glucose-induced cardiac fibrosis through the JNK/SAPK and p38 signalling pathways by suppressing DUSP8 expression.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder