Merck
Cadmium-induced genotoxicity in human osteoblast-like cells.
Mutation research. Genetic toxicology and environmental mutagenesis (2014-12-02)
Helena Oliveira, Cristina Monteiro, Francisco Pinho, Sónia Pinho, José Miguel P Ferreira de Oliveira, Conceição Santos
ABSTRACT
Cadmium (Cd) is a widespread heavy metal used in numerous industrial processes. Cd exerts toxicological effects mostly in kidney and liver. Bone is also an important target of Cd, however, the cellular mechanisms of Cd toxicological effects in the bone cells are still poorly understood. Therefore, the present work aimed to investigate the putative cytotoxic and genotoxic effects of Cd to human bone cells. For that, the osteoblast-like MG-63 cells were exposed to 20 and 50μM Cd for 24 and 48h. Results showed a dose-dependent increase in Cd accumulation in cells and a decrease in cell viability, especially after 48h. Cell cycle analysis showed a delay at S phase concomitant with a decrease in cells at G0/G1 phase. After 24h, Cd treatment downregulated the expression of CHEK1, CHEK2 and CDK2 genes and upregulated the expression of CCNE1 gene. After 48h, the expression of ATM and CCNB1 genes were downregulated. Also, a 3.3 fold increase on the expression of gene CCNE1 was detected. Both Cd doses induced DNA fragmentation at 48h, while an increase in micronuclei (MN) and nucleoplasmic bridges (NPBs) together with an increase in the percentage of apoptotic/necrotic cells was detected for both time periods. Overall, our results demonstrate the cytotoxicity and genotoxicity of Cd in human bone cells. Also, the cytokinesis-block micronucleus (CBMN) assay parameters (MN, NPBs and the percentage of cells under apoptosis or necrosis) together with the cell cycle appear as the most sensitive to Cd cyto- and genotoxicity, being early affected even with the lowest Cd dose. Therefore, these cyto-/genotoxic techniques may be selected for early detection of Cd-induced toxicity.
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, wire reel, 0.1m, diameter 1.0mm, hard, 99.999%
Cadmium, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, wire reel, 0.5m, diameter 0.5mm, hard, 99.95+%
Cadmium, foil, 25mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, light tested, 50x50mm, thickness 0.025mm, 99.7+%
Cadmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, 6mm disks, thickness 0.005mm, 99.7+%
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cadmium, foil, not light tested, 25x25mm, thickness 0.002mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, light tested, 50x50mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.01mm, 99.7+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 6mm disks, thickness 0.015mm, 99.7+%
Cadmium, rod, 50mm, diameter 40mm, 99.9%
Cadmium, foil, 4mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, light tested, 25x25mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%