Merck
  • Palmitate induces insulin resistance in human HepG2 hepatocytes by enhancing ubiquitination and proteasomal degradation of key insulin signaling molecules.
Palmitate induces insulin resistance in human HepG2 hepatocytes by enhancing ubiquitination and proteasomal degradation of key insulin signaling molecules.
Archives of biochemistry and biophysics (2014-12-21)
Megumi Ishii, Ayumi Maeda, Shuji Tani, Mitsugu Akagawa
ABSTRACT
Obesity-associated insulin resistance is a major pathogenesis of type 2 diabetes mellitus and is characterized by defects in insulin signaling. High concentrations of plasma free fatty acids (FFAs) are involved in the etiology of obesity-associated insulin resistance. However, the detailed mechanism by which FFAs contribute to the development of insulin resistance is not yet fully understood. We investigated the molecular basis of insulin resistance elicited by FFAs using the human hepatocyte cell line HepG2. Among major human FFAs, palmitate markedly inhibited insulin-stimulated phosphorylation of key insulin signaling molecules such as insulin receptor, insulin receptor substrate-1, and Akt, indicating that palmitate is the principal inducer of insulin resistance. We revealed that palmitate facilitates ubiquitination of the key insulin signaling molecules, and subsequently elicits their proteasomal degradation. Furthermore, we demonstrated that inhibition of ubiquitination by the ubiquitin-activating enzyme E1 inhibitor PYR41 significantly prevents palmitate-inducible insulin resistance but not by the proteasome inhibitor MG132, implying that ubiquitinated signaling molecules may be dysfunctional. In conclusion, inhibition of ubiquitination of the key insulin signaling molecules may be a potential strategy for preventing and treating obesity-associated insulin resistance.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Palmitic acid, ≥99%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified