Merck
  • Diagnosis and treatment of diffuse large B-cell lymphoma in an orangutan (Pongo pygmaeus).
Diagnosis and treatment of diffuse large B-cell lymphoma in an orangutan (Pongo pygmaeus).
Journal of zoo and wildlife medicine : official publication of the American Association of Zoo Veterinarians (2015-01-31)
Offiong F Ikpatt, Drury Reavill, Jason Chatfield, Susan Clubb, Joseph D Rosenblatt, Glenn Fonte, Yao-Shan Fan, Carolyn Cray
ABSTRACT
Lymphoma is a common malignancy observed in companion animals. This type of naturally occurring neoplasia has been uncommonly reported in great apes. Diffuse large B-cell lymphoma was diagnosed in an 8-yr-old captive orangutan (Pongo pygmaeus) with gastrointestinal disease by histologic and immunohistochemical methodologies. The orangutan was treated with three cycles of combination chemotherapy (intravenous Rituxan, cyclophosphamide, doxorubicin, and vincristine). The primate has been in good health and exhibiting normal behaviors for more than 15 mo following treatment.
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Prednisone, ≥98%
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard