Merck
A role for resistin in the ovary during the estrous cycle.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme (2014-03-15)
A Rak-Mardyła, M Duda, E Ł Gregoraszczuk
ABSTRACT
In a previous study, we showed that resistin expression increased during ovarian follicle development in prepubertal pigs and had direct effects on steroidogenesis, suggesting an important role for resistin in the ovary during puberty. To determine its potential regulatory role in the ovary during the estrous cycle, using real-time PCR, immunoblotting, immunohistochemistry, and ELISA methods, we quantified the expression, immunolocalization and concentration of resistin in different sized ovarian follicles (small, 2-4 mm; medium, 4-6 mm; and large, 8-12 mm) in mature pigs. We then determined the effects of recombinant resistin (0.1, 1, and 10 ng/ml) on steroid hormone (progesterone-P4, androstendione-A4, testosterone-T, and estradiol-E2) secretion and steroidogenic enzyme (3βHSD, CYP17A1, 17βHSD, and CYP19A1) gene and protein expression in ovarian follicles. We found no differences in the resistin expression between all of the examined follicles. Immunostaining analysis also showed resistin expression in the cytoplasm of both granulosa and theca cells, where it was localized more abundantly in the granulosa cells compared to the theca cells. Recombinant resistin direct stimulated P4, A4, and T secretion via increased expression of 3βHSD, CYP17A1, and 17βHSD, suggesting an autocrine and/or paracrine regulatory role in the porcine ovary during the estrous cycle.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)