Merck
  • Heat-shock protein A8 restores sperm membrane integrity by increasing plasma membrane fluidity.
Heat-shock protein A8 restores sperm membrane integrity by increasing plasma membrane fluidity.
Reproduction (Cambridge, England) (2014-02-07)
Najmeh Moein-Vaziri, Ian Phillips, Simon Smith, Carmen Almiňana, Carolina Maside, Maria A Gil, Jordi Roca, Emilio A Martinez, William V Holt, A Graham Pockley, Alireza Fazeli
ABSTRACT
The constitutive 70  kDa heat-shock protein, HSPA8, has previously been shown to contribute to the long-term survival of spermatozoa inside the mammalian female reproductive tract. Here, we show that a recombinant form of HSPA8 rapidly promotes the viability of uncapacitated spermatozoa, the ability of spermatozoa to bind to oviductal epithelial cells, enhances IVF performance, and decreases sperm mitochondrial activity. Fluorescence recovery after photobleaching revealed that the repair of membrane damage is achieved by an almost instantaneous increase in sperm membrane fluidity. The ability of HSPA8 to influence membrane stability and fluidity, as well as its conserved nature among mammalian species, supports the idea that this protein protects sperm survival through membrane repair mechanisms. Free Persian abstract A Persian translation of the abstract is freely available online at http://www.reproduction-online.org/content/147/5/719/suppl/DC1.
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Ethidium homodimer, suitable for fluorescence, ~90% (HPCE)
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Urea, European Pharmacopoeia (EP) Reference Standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
Urea, analytical standard
Sigma-Aldrich
Anti-HSPA8 antibody produced in rabbit