Merck
  • Simple and efficient methods for enrichment and isolation of endonuclease modified cells.
Simple and efficient methods for enrichment and isolation of endonuclease modified cells.
PloS one (2014-05-07)
Branden S Moriarity, Eric P Rahrmann, Dominic A Beckmann, Caitlin B Conboy, Adrienne L Watson, Daniel F Carlson, Erik R Olson, Kendra A Hyland, Scott C Fahrenkrug, R Scott McIvor, David A Largaespada
ABSTRACT
The advent of Transcription Activator-Like Effector Nucleases (TALENs), and similar technologies such as CRISPR, provide a straightforward and cost effective option for targeted gene knockout (KO). Yet, there is still a need for methods that allow for enrichment and isolation of modified cells for genetic studies and therapeutics based on gene modified human cells. We have developed and validated two methods for simple enrichment and isolation of single or multiplex gene KO's in transformed, immortalized, and human progenitor cells. These methods rely on selection of a phenotypic change such as resistance to a particular drug or ability to grow in a selective environment. The first method, termed co-transposition, utilizes integration of a piggyBac transposon vector encoding a drug resistance gene. The second method, termed co-targeting, utilizes TALENs to KO any gene that when lost induces a selectable phenotype. Using these methods we also show removal of entire genes and demonstrate that TALENs function in human CD34+ progenitor cells. Further, co-transposition can be used to generate conditional KO cell lines utilizing an inducible cDNA rescue transposon vector. These methods allow for robust enrichment and isolation of KO cells in a rapid and efficient manner.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Dipyridamole, ≥98% (HPLC)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Dipyridamole, European Pharmacopoeia (EP) Reference Standard
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
6-Thioguanine, ≥98%
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
6-Thioguanine, Hybri-Max, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
SAFC
Methotrexate
Dipyridamole for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Thioguanine, United States Pharmacopeia (USP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material