Merck
  • Mesenchymal stromal cell proliferation, gene expression and protein production in human platelet-rich plasma-supplemented media.
Mesenchymal stromal cell proliferation, gene expression and protein production in human platelet-rich plasma-supplemented media.
PloS one (2014-08-15)
Paola Romina Amable, Marcus Vinicius Telles Teixeira, Rosana Bizon Vieira Carias, José Mauro Granjeiro, Radovan Borojevic
ABSTRACT
Platelet-rich plasma (PRP) is increasingly used as a cell culture supplement, in order to reduce the contact of human cells with animal-derived products during in vitro expansion. The effect of supplementation changes on cell growth and protein production is not fully characterized. Human mesenchymal stromal cells from bone marrow, adipose tissue and Wharton's Jelly were isolated and cultured in PRP-supplemented media. Proliferation, in vitro differentiation, expression of cell surface markers, mRNA expression of key genes and protein secretion were quantified. 10% PRP sustained five to tenfold increased cell proliferation as compared to 10% fetal bovine serum. Regarding cell differentiation, PRP reduced adipogenic differentiation and increased calcium deposits in bone marrow and adipose tissue-mesenchymal stromal cells. Wharton's Jelly derived mesenchymal stromal cells secreted higher concentrations of chemokines and growth factors than other mesenchymal stromal cells when cultured in PRP-supplemented media. Bone marrow derived mesenchymal stromal cells secreted higher concentrations of pro-inflammatory and pro-angiogenic proteins. Mesenchymal stromal cells isolated from adipose tissue secreted higher amounts of extracellular matrix components. Mesenchymal stromal cells purified from different tissues have distinct properties regarding differentiation, angiogenic, inflammatory and matrix remodeling potential when cultured in PRP supplemented media. These abilities should be further characterized in order to choose the best protocols for their therapeutic use.
Product Description
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Transferrin human, powder, BioReagent, suitable for cell culture
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
TGF-beta 3-E. coli human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Alizarin Red S, certified by the BSC
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture