- Formation and characteristics of Cu colloids from CuO powder by laser irradiation in 2-propanol.
Formation and characteristics of Cu colloids from CuO powder by laser irradiation in 2-propanol.
The Journal of Physical Chemistry B (1999)
Yeh MS, et al.