Merck
  • Improving the odorant sensitivity of olfactory receptor-expressing yeast with accessory proteins.
Improving the odorant sensitivity of olfactory receptor-expressing yeast with accessory proteins.
Analytical biochemistry (2014-12-03)
Yosuke Fukutani, Ayaka Hori, Satoshi Tsukada, Ryoichi Sato, Jun Ishii, Akihiko Kondo, Hiroaki Matsunami, Masafumi Yohda
ABSTRACT
Olfaction depends on the selectivity and sensitivity of olfactory receptors. Previous attempts at constructing a mammalian olfactory receptor-based artificial odorant sensing system in the budding yeast Saccharomyces cerevisiae suffered from low sensitivity and activity. This result may be at least in part due to poor functional expression of olfactory receptors and/or limited solubility of some odorants in the medium. In this study, we examined the effects of two types of accessory proteins, receptor transporting protein 1 short and odorant binding proteins, in improving odor-mediated activation of olfactory receptors expressed in yeast. We found that receptor transporting protein 1 short enhanced the membrane expression and ligand-induced responses of some olfactory receptors. Coexpression of odorant binding proteins of the silkworm moth Bombyx mori enhanced the sensitivity of a mouse olfactory receptor. Our results suggest that different classes of accessory proteins can confer sensitive and robust responses of olfactory receptors expressed in yeast. Inclusion of accessory proteins may be essential in the future development of practical olfactory receptor-based odorant sensors.
Product Description
Supelco
Eugenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
5α-Androst-16-en-3-one
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetophenone, analytical standard
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Supelco
Acetophenone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Eugenol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetophenone, natural, 98%, FG