Merck
Use of human serum for human corneal endothelial cell culture.
The British journal of ophthalmology (2014-12-03)
Lucas Monferrari Monteiro Vianna, Laura Kallay, Tetsuya Toyono, Rubens Belfort, Jeffrey D Holiman, Albert S Jun
ABSTRACT
To study human corneal endothelial cells (HCECs) cultured in vitro with human serum (HS) supplemented media (HS-SM) compared with HCEC cultured in fetal bovine serum (FBS) supplemented media (FBS-SM). One cornea from a donor aged 21 years and a pair of corneas from a 16 year-old donor were obtained from the eye bank and used to create two different cell populations. At the first passage, the cell populations were equally divided and seeded in two different wells containing FBS-SM or HS-SM. In subsequent passages, HS-SM was compared with FBS-SM by morphology, growth curves, immunohistochemistry and real-time reverse-transcriptase PCR for endothelial cell markers. No difference in morphology could be seen in P2, P5 or in any other passages for cells grown in the two media. By growth curves, cell counts were similar in FBS-SM and HS-SM from days 1 to 5, with a trend towards higher cell counts in HS-SM at day 7. Cells grown in FBS-SM and HS-SM media showed similar expression of endothelial cell markers when assessed by immunohistochemistry and real-time reverse-transcriptase PCR. HS-SM was similar to FBS-SM for HCEC culture when assessed by cell morphology, proliferation and protein/gene expression.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis