Merck
  • The luteinising hormone surge-generating system is functional in male goats as in females: involvement of kisspeptin neurones in the medial preoptic area.
The luteinising hormone surge-generating system is functional in male goats as in females: involvement of kisspeptin neurones in the medial preoptic area.
Journal of neuroendocrinology (2014-11-05)
F Matsuda, K Nakatsukasa, Y Suetomi, Y Naniwa, D Ito, N Inoue, Y Wakabayashi, H Okamura, K-I Maeda, Y Uenoyama, H Tsukamura, S Ohkura
ABSTRACT
A luteinising hormone (LH) surge is fundamental to the induction of ovulation in mammalian females. The administration of a preovulatory level of oestrogen evokes an LH surge in ovariectomised females, whereas the response to oestrogen in castrated males differs among species; namely, the LH surge-generating system is sexually differentiated in some species (e.g. rodents and sheep) but not in others (e.g. primates). In the present study, we aimed to determine whether there is a functional LH surge-generating system in male goats, and whether hypothalamic kisspeptin neurones in male goats are involved in the regulation of surge-like LH secretion. By i.v. infusion of oestradiol (E2; 6 μg/h) for 16 h, a surge-like LH increase occurred in both castrated male and ovariectomised female goats, although the mean peak LH concentration was lower and the mean peak of the LH surge was later in males compared to females. Dual staining with KISS1 in situ hybridisation and c-Fos immunohistochemistry revealed that E2 treatment significantly increased c-Fos expression in the medial preoptic area (mPOA) KISS1 cells in castrated males, as well as ovariectomised females. By contrast, dual-labelled cells were scarcely detected in the arcuate nucleus (ARC) after E2 treatment in both sexes. These data suggest that kisspeptin neurones in the mPOA, but not those in the ARC, are involved in the induction of surge-like LH secretion in both male and female goats. In summary, our data show that the mechanism that initiates the LH surge in response to oestrogen, the mPOA kisspeptin neurones, is functional in male goats. Thus, sexual differentiation of the LH surge-generating system would not be applicable to goats.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Trolamine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Tetrazolium Blue Chloride, used in colorimetric determination of reducing compounds
Sigma-Aldrich
Tetrazolium Blue Chloride, suitable for microbiology, ≥90% (T)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Triethanolamine, analytical standard
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)