Merck
  • Soybean chalcone isomerase: evolution of the fold, and the differential expression and localization of the gene family.
Soybean chalcone isomerase: evolution of the fold, and the differential expression and localization of the gene family.
Planta (2014-11-12)
Mehran Dastmalchi, Sangeeta Dhaubhadel
ABSTRACT
Soybean chalcone isomerase (CHI) family contains twelve members with unique evolutionary background, expression patterns and is compartmentalized to specific subcellular locations. The phenylpropanoid pathway produces a diverse array of plant natural products. A key branch-point enzyme, chalcone isomerase, catalyzes the reaction producing flavanones, the backbone for many downstream metabolites such as flavonoids and isoflavonoids. We have identified twelve soybean GmCHIs that fall into four subfamilies. The study of this family in soybean in the context of various CHIs and CHI-like proteins, across divisions in the plant kingdom and beyond, shows an evolutionary journey from fatty acid-binding proteins (FAPs) to sterically restricted folds that gave rise to the chalcone-to-flavanone isomerase. There are four GmCHIs with this functionality, three of which belong to a legume-specific clade known as 'type II' CHIs. Tissue-specific expression of eight core members of the soybean CHI family showed differential temporal and spatial expression, pointing to the potential function of GmCHI1A in seed isoflavonoid production. Promoter analysis of the GmCHIs described the minutiae of sub-organ expression patterns. Subcellular localization of the family was conducted to investigate the possibility of pathway-specific compartmentalization. Subfamilies 1, 2 and 4 localized to the nucleus and cytoplasm, with nuclear localization of CHIs raising questions about alternate function. GmCHI3 isoforms localized to the chloroplast, which, in conjunction with their position on the phylogenetic tree and expression patterns, closely associates them with the FAPs. This study provides the first comprehensive look at soybean CHIs, a family of unique evolutionary background and biochemical function, with the catalytically active members producing the backbone substrate in an important plant metabolic pathway.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard