Merck
  • The hydrolysed products of iridoid glycosides can enhance imatinib mesylate-induced apoptosis in human myeloid leukaemia cells.
The hydrolysed products of iridoid glycosides can enhance imatinib mesylate-induced apoptosis in human myeloid leukaemia cells.
Phytotherapy research : PTR (2015-01-15)
Min-Beom Kim, Chulwon Kim, Won-Seok Chung, Jae-Heung Cho, Dongwoo Nam, Sung-Hoon Kim, Kwang Seok Ahn
ABSTRACT
Several studies have demonstrated that deregulated activation of signal transducer and activator of transcription 3 (STAT3) has been associated with survival, proliferation, chemoresistance and angiogenesis of tumour cells. Thus, inhibition of STAT3 expression could be a potent therapeutic approach for cancer treatment. Using several leukaemia cell lines, the effect of the hydrolysed-catalpol (H-catalpol) and hydrolysed-aucubin (H-aucubin) products on the STAT3 signalling pathway, inhibition of BCR-ABL activation, cellular proliferation and potentiation of imatinib mesylate-induced apoptosis was investigated. We found that iridoid glycosides (catalpol and aucubin) did not exert any cytotoxicity in the tumour cells, whereas both H-catalpol and H-aucubin exhibited significant cytotoxic effects on K562 human myeloid leukaemia cells. Indeed, H-catalpol and H-aucubin down-regulated BCR-ABL phosphorylation and inhibited constitutive STAT3 activation through abrogating upstream JAK2 and c-Src and constitutive STAT5 activation leading to apoptosis through caspase-3 activation. Hydrolysed-catalpol enhanced the apoptosis induced by imatinib mesylate and this correlated with down-regulation of gene products that mediate cell proliferation (cyclin D1), and cell survival (Bcl-2, Bcl-xL and survivin); all known to be regulated by the STAT3. Overall, our results provide novel insight into the role of hydrolysed iridoids in potentially treating leukaemia through the modulation of STAT3 signalling pathway.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
SAFC
Tromethamine
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride, tablet
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture