Merck
  • Development of an equilibrium headspace gas chromatographic method for the measurement of noncovalent association and partitioning of n-alkylbenzenes at infinite dilution in fulvic acid pseudophase.
Development of an equilibrium headspace gas chromatographic method for the measurement of noncovalent association and partitioning of n-alkylbenzenes at infinite dilution in fulvic acid pseudophase.
Journal of chromatography. A (2015-01-28)
Mahmoud D Eljack, Rachael E Wilson, Abul Hussam, Shahamat U Khan
ABSTRACT
Fulvic acid (FA), the most important water soluble fraction of humic substances in nature, is known to form aggregate pseudophase and complexes with organic and inorganic species. Here, we report a novel equilibrium headspace gas chromatography (eHSGC) and a two-step reaction model to measure n-alkylbenzene-FA association constant (K11) and n-alkylbenzene-pseudophase FAn association constant (Kn1) without solute concentration and response factor. The K11 and Kn1 values were 2-3 orders of magnitude higher than those for sodium dodecylsulfate. Changes in peak area were used to calculate the critical FA-aggregation concentration (cfc), mole fraction based partition coefficients (Kx), activity coefficients of solute inside the aggregate pseudophase (γm(∞)), and transfer free energies of alkyl CH2 at infinite dilution. The cfc was found to be 10±0.5μM. The Kx values are of the order of 10(7) in the FA-aggregate pseudophase. The data shows that benzene has the lowest (0.0002) and n-butylbenzene has the highest (0.01) γm(∞) values, which are seven orders of magnitude smaller than γw(∞) in water. The transfer free energy of association of a CH2 group, -155cal/mol, compared to that of benzene, -9722cal/mol, indicates that the FA-aggregate pseudophase is more polarizable benzene-like and less n-alkane aliphatic-like.
Sigma-Aldrich
Rhein
Sigma-Aldrich
Rhein, technical grade
Supelco
Butylbenzene, analytical standard
Sigma-Aldrich
Butylbenzene, ≥99%
Sigma-Aldrich
Propylbenzene, 98%
Sigma-Aldrich
Propylbenzene, ≥99.0% (GC)
Supelco
Propylbenzene, analytical standard
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Supelco
Ethylbenzene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Toluene, analytical standard