Merck
  • Practical approach to prepare solid dispersion drug product using spherical silicate.
Practical approach to prepare solid dispersion drug product using spherical silicate.
International journal of pharmaceutics (2014-08-19)
Kentaro Nagane, Susumu Kimura, Koji Ukai, Noriko Ogawa, Hiromitsu Yamamoto
ABSTRACT
The purpose of this study is to establish a novel approach for preparing a solid dispersion drug product using spherical silicate by a Wurster-type fluidized bed granulator. The spherical silicate used in this study has porous structure and ideal particle size for loading by a Wurster-type fluidized bed granulator. As model drugs, ibuprofen (IBU), indomethacin (IMC), and phenytoin (PNT) were used and the proposed approach was applied to prepare amorphous drug. All drugs could be loaded on the spherical silicate in an amorphous state. On the other hand, spray drying of spherical silicate suspended in IBU solution was conducted to prepare amorphous product of IBU as a reference; however, complete amorphization was not achieved. Dissolution profiles of each drug after loading on spherical silicate by a Wurster-type fluidized bed granulator were evaluated, and dramatic improvement of dissolution was observed compared with those of crystalline drug. In the proposed approach, specific surface area and particle size of spherical silicate were determined as a key factors to contribute to high yield of amorphous product.
Product Description
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Indomethacin, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Supelco
Ibuprofen
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Phenytoin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
5,5-Diphenylhydantoin, ≥98%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Phenytoin, European Pharmacopoeia (EP) Reference Standard
Phenytoin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications