Merck
  • Abundance of live ²⁴⁴Pu in deep-sea reservoirs on Earth points to rarity of actinide nucleosynthesis.
Abundance of live ²⁴⁴Pu in deep-sea reservoirs on Earth points to rarity of actinide nucleosynthesis.
Nature communications (2015-01-21)
A Wallner, T Faestermann, J Feige, C Feldstein, K Knie, G Korschinek, W Kutschera, A Ofan, M Paul, F Quinto, G Rugel, P Steier
ABSTRACT
Half of the heavy elements including all actinides are produced in r-process nucleosynthesis, whose sites and history remain a mystery. If continuously produced, the Interstellar Medium is expected to build-up a quasi-steady state of abundances of short-lived nuclides (with half-lives ≤100 My), including actinides produced in r-process nucleosynthesis. Their existence in today's interstellar medium would serve as a radioactive clock and would establish that their production was recent. In particular (244)Pu, a radioactive actinide nuclide (half-life=81 My), can place strong constraints on recent r-process frequency and production yield. Here we report the detection of live interstellar (244)Pu, archived in Earth's deep-sea floor during the last 25 My, at abundances lower than expected from continuous production in the Galaxy by about 2 orders of magnitude. This large discrepancy may signal a rarity of actinide r-process nucleosynthesis sites, compatible with neutron-star mergers or with a small subset of actinide-producing supernovae.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 7 N in methanol