Merck
  • High expression of miR-21 in triple-negative breast cancers was correlated with a poor prognosis and promoted tumor cell in vitro proliferation.
High expression of miR-21 in triple-negative breast cancers was correlated with a poor prognosis and promoted tumor cell in vitro proliferation.
Medical oncology (Northwood, London, England) (2014-06-16)
Guizhi Dong, Xiaoling Liang, Deguang Wang, Huiquan Gao, Ling Wang, Lili Wang, Jingjun Liu, Zhaohui Du
ABSTRACT
The triple-negative breast cancer (TNBC), with a particularly poor prognosis, is increasingly recognized as heterogeneous in molecular signatures. MicroRNA expression profiles have been used for the classification and prognostication of breast cancer, numerous significantly upregulated microRNAs, i.e. miR-21, have been verified oncogenic in non-TNBCs. In present study, we determined the miR-21 levels in TNBC specimens, and TNBC cell levels in vitro, and then identified the role of miR-21 on tumor cell proliferation, apoptosis, and then identified PTEN as the possible target of the microRNA. It was shown that miR-21 expression is upregulated generally, and heterogeneous in TNBC specimens, posing a correlation with poor prognosis for TNBC patients. Further results demonstrated that the upregulated miR-21 promoted the tumor proliferation and inhibited cell apoptosis in vitro. And pro-apoptotic PTEN had been shown being targeted and downregulated. Therefore, our finding emphasized the oncogenic role of miR-21 in TNBC.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Supelco
5-Fluorouracil, analytical standard
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Monoclonal Anti-CASP9 antibody produced in mouse, clone 3B8-4G2, purified immunoglobulin, buffered aqueous solution
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard