Merck
  • Facile enzymatic synthesis of sugar 1-phosphates as substrates for phosphorylases using anomeric kinases.
Facile enzymatic synthesis of sugar 1-phosphates as substrates for phosphorylases using anomeric kinases.
Carbohydrate research (2014-12-03)
Yuan Liu, Mamoru Nishimoto, Motomitsu Kitaoka
ABSTRACT
Three sugar 1-phosphates that are donor substrates for phosphorylases were produced at the gram scale from phosphoenolpyruvic acid and the corresponding sugars by the combined action of pyruvate kinase and the corresponding anomeric kinases in good yields. These sugar 1-phosphates were purified through two electrodialysis steps. α-D-Galactose 1-phosphate was finally isolated as crystals of dipotassium salts. α-D-Mannose 1-phosphate and 2-acetamido-2-deoxy-α-D-glucose 1-phosphate were isolated as crystals of bis(cyclohexylammonium) salts.
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of 30 mg
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade II, ≥97% (HPLC), crystalline, from microbial
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial