Merck
  • Phosphorylation of glucocorticoid receptor tau1c transactivation domain enhances binding to CREB binding protein (CBP) TAZ2.
Phosphorylation of glucocorticoid receptor tau1c transactivation domain enhances binding to CREB binding protein (CBP) TAZ2.
Biochemical and biophysical research communications (2014-12-17)
Carl W Carruthers, Ji Ho Suh, Jan-Ake Gustafsson, Paul Webb
ABSTRACT
The glucocorticoid receptor (GR) N-terminal domain (NTD) contains a transactivation domain (activation function 1; AF-1). GR AF-1 is phosphorylated, but effects of this modification upon AF-1 activity and cofactor recruitment are not completely clear. GR AF-1 activity is mostly confined to a short unstructured domain called tau1c (amino acids 187-244) that contains three phosphorylation sites and binds a short cysteine rich fragment (CH3) of the coactivator CREB binding protein (CBP). Since the CH3 domain overlaps the CBP transcriptional adaptor zinc binding (TAZ) 2 domain, implicated in phosphorylation dependent binding to other unstructured transcription factor domains, we set out to investigate whether GR interacts with TAZ2 and whether this binding event is modulated by phosphorylation. We find that GR tau1c is absolutely required for enhancement of GR function and GR/CBP association in cultured cells. Tau1c interacts with TAZ2 in vitro and peptide mapping reveals CBP binding determinants throughout tau1c. Phosphorylation at GR Ser203, not involved in transactivation, does not affect tau1c/TAZ2 interactions. However, phosphorylation at Ser211 and Ser226, markers of GR transcriptional activity, greatly enhances TAZ2 binding in a synergistic fashion. We propose that GR tau1c phosphorylation could promote CBP recruitment and enhance AF-1 activity.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard