Merck
  • Rapamycin improves peripheral nerve myelination while it fails to benefit neuromuscular performance in neuropathic mice.
Rapamycin improves peripheral nerve myelination while it fails to benefit neuromuscular performance in neuropathic mice.
Neurobiology of disease (2014-07-12)
Jessica Nicks, Sooyeon Lee, Andrew Harris, Darin J Falk, Adrian G Todd, Karla Arredondo, William A Dunn, Lucia Notterpek
ABSTRACT
Charcot--Marie-Tooth disease type 1A (CMT1A) is a hereditary peripheral neuropathy characterized by progressive demyelination and distal muscle weakness. Abnormal expression of peripheral myelin protein 22 (PMP22) has been linked to CMT1A and is modeled by Trembler J (TrJ) mice, which carry the same leucine to proline substitution in PMP22 as affected pedigrees. Pharmacologic modulation of autophagy by rapamycin in neuron-Schwann cell explant cultures from neuropathic mice reduced PMP22 aggregate formation and improved myelination. Here we asked whether rapamycin administration by food supplementation, or intraperitoneal injection, could alleviate the neuropathic phenotype of affected mice and improve neuromuscular performance. Cohorts of male and female wild type (Wt) and TrJ mice were assigned to placebo or rapamycin treatment starting at 2 or 4months of age and tested monthly on the rotarod. While neither long-term feeding (8 or 10months) on rapamycin-enriched diet, or short-term injection (2months) of rapamycin improved locomotor performance of the neuropathic mice, both regimen benefited peripheral nerve myelination. Together, these results indicate that while treatment with rapamycin benefits the myelination capacity of neuropathic Schwann cells, this intervention does not improve neuromuscular function. The observed outcome might be the result of the differential response of nerve and skeletal muscle tissue to rapamycin.
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Glycerol, FCC, FG
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)