Merck
  • Thermal degradation of green tea flavan-3-ols and formation of hetero- and homocatechin dimers in model dairy beverages.
Thermal degradation of green tea flavan-3-ols and formation of hetero- and homocatechin dimers in model dairy beverages.
Food chemistry (2014-12-04)
Brian J Song, Chris Manganais, Mario G Ferruzzi
ABSTRACT
Interactions between polyphenols and macromolecules may impact polyphenol stability and bioavailability from foods. The impact of milk on tea flavan-3-ol stability to thermal treatment was investigated. Single strength (36.2 protein per L), quarter strength (9.0 g protein per L) milk, and control model beverages were incubated with epigallocatechin gallate and green tea extract at 62 or 37 °C for 180 min. Intact flavan-3-ols and select auto-oxidation products [theasinesins (THSNs) and P-2 dimers] were quantified by LC-MS. Generally, greater polyphenol to protein ratios increased first order degradation rates, consequently decreasing formation of oxidation products. The presence of galloyl and hydroxy moieties was associated with higher stability of monomeric flavan-3-ols with increasing protein concentrations suggesting potential for protein affinity to stabilise flavan-3-ols to thermal treatment. Absence of these moieties led to no observable improvements in stability. These results suggest that protein interactions may be useful in stabilising flavan-3-ols through thermal processing.
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Supelco
Methanol, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%