Merck
  • Target-controlled infusion and population pharmacokinetics of landiolol hydrochloride in gynecologic patients.
Target-controlled infusion and population pharmacokinetics of landiolol hydrochloride in gynecologic patients.
Journal of anesthesia (2014-09-05)
Takayuki Kunisawa, Akio Yamagishi, Manabu Suno, Susumu Nakade, Ryunosuke Higashi, Atsushi Kurosawa, Ami Sugawara, Kazuo Matsubara, Hiroshi Iwasaki
ABSTRACT
We previously determined the pharmacokinetic (PK) parameters of landiolol in healthy male volunteers. In this study, we evaluated the usefulness of target-controlled infusion (TCI) of landiolol hydrochloride and determined PK parameters of landiolol in gynecologic patients. Nine patients who were scheduled to undergo gynecologic surgery were enrolled. After inducing anesthesia, landiolol hydrochloride was administered at the target plasma concentrations of 500 and 1,000 ng/mL for each 30 min. A total of 126 data points of plasma concentration were collected from the patients and used for the population PK analysis. Furthermore, a population PK model was developed using the nonlinear mixed-effect modeling software. The patients had markedly decreased heart rates (HRs) at 2 min after the initiation of landiolol hydrochloride administration; however, their blood pressures did not markedly change from the baseline value. The concentration time course of landiolol was best described by a 2-compartment model with lag time. The estimate of PK parameters were total body clearance (CL) 34.0 mL/min/kg, distribution volume of the central compartment (V 1) 74.9 mL/kg, inter-compartmental clearance (Q) 70.9 mL/min/kg, distribution volume of the peripheral compartment (V 2) 38.9 mL/kg, and lag time (ALAG) 0.634 min. The predictive performance of this model was better than that of the previous model. TCI of landiolol hydrochloride is useful for controlling HR, and the PK parameters of landiolol in gynecologic patients were similar to those in healthy male volunteers and best described by a 2-compartment model with lag time.
Product Description
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Propofol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Propofol for peak identification, European Pharmacopoeia (EP) Reference Standard