Merck
  • Effect of selective inhibition of monoacylglycerol lipase (MAGL) on acute nausea, anticipatory nausea, and vomiting in rats and Suncus murinus.
Effect of selective inhibition of monoacylglycerol lipase (MAGL) on acute nausea, anticipatory nausea, and vomiting in rats and Suncus murinus.
Psychopharmacology (2014-08-03)
Linda A Parker, Micah J Niphakis, Rachel Downey, Cheryl L Limebeer, Erin M Rock, Martin A Sticht, Heather Morris, Rehab A Abdullah, Aron H Lichtman, Benjamin F Cravatt
ABSTRACT
To determine the role of the endocannabinoid, 2-arachodonyl glycerol (2-AG), in the regulation of nausea and vomiting. We evaluated the effectiveness of the potent selective monoacylglycerol lipase (MAGL) inhibitor, MJN110, which selectively elevates the endocannabinoid 2-AG, to suppress acute nausea and vomiting, as well as anticipatory nausea in rat and shrew models. The rat gaping models were used to evaluate the potential of MJN110 (5, 10, and 20 mg/kg, intraperitoneally [IP]) to suppress acute nausea produced by LiCl and of MJN110 (10 and 20 mg/kg, IP) to suppress anticipatory nausea elicited by a LiCl-paired context. The potential as well of MJN110 (10 and 20 mg/kg, IP) to suppress vomiting and contextually elicited gaping in the Suncus murinus was evaluated. MJN110 suppressed acute nausea in rats, LiCl-induced vomiting in shrews and contextually-elicited anticipatory nausea in both rats (accompanied by elevation of 2-AG in the visceral insular cortex) and shrews. These effects were reversed by the CB1 antagonist/inverse agonist, SR141716. The MAGL inhibitor did not modify locomotion at any dose. An activity-based protein profiling analysis of samples of tissue collected from the visceral insular cortex in rats and whole brain tissues in shrews revealed that MJN110 selectively inhibited MAGL and the alternative 2-AG hydrolase, ABHD6. MAGL inhibition by MJN110 which selectively elevates endogenous 2-AG has therapeutic potential in the treatment of acute nausea and vomiting as well as anticipatory nausea, a distressful symptom that is resistant to currently available treatments.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Saccharin, ≥99%
Saccharin, European Pharmacopoeia (EP) Reference Standard
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
2-Arachidonyl glycerol, ~10 mg/mL, ≥98% (HPLC)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
MJN110, ≥98% (HPLC)
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Lithium chloride solution, 2 M in ethanol
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol