Merck
  • Intravenous immunoglobulin attenuates experimental autoimmune arthritis by inducing reciprocal regulation of Th17 and Treg cells in an interleukin-10-dependent manner.
Intravenous immunoglobulin attenuates experimental autoimmune arthritis by inducing reciprocal regulation of Th17 and Treg cells in an interleukin-10-dependent manner.
Arthritis & rheumatology (Hoboken, N.J.) (2014-03-20)
Seon-Yeong Lee, Young-Ok Jung, Jun-Geol Ryu, Chang-Min Kang, Eun-Kyung Kim, Hye-Jin Son, Eun-Ji Yang, Ji-Hyeon Ju, Young-Sun Kang, Sung-Hwan Park, Ho-Youn Kim, Mi-La Cho
ABSTRACT
Intravenous immunoglobulin (IVIG) is used as a therapeutic agent in various autoimmune diseases. The aims of this study were to investigate the therapeutic effects of IVIG on collagen-induced arthritis (CIA) and identify the mechanism responsible for any therapeutic effects. IVIG was administered to mice with CIA, and the in vivo effects were determined. Th17 and Treg cell frequencies were analyzed by flow cytometry, and cytokine levels in the supernatant were measured by enzyme-linked immunosorbent assay. Subpopulations of T cells and B cells in the spleen were assessed by confocal microscopy. The arthritis severity score and incidence of arthritis were lower in mice treated with IVIG compared with untreated mice. Histopathologic analysis showed less joint damage in mice treated with IVIG. The expression of proinflammatory cytokines, specific type II collagen antibodies, and osteoclast markers was significantly reduced in mice treated with IVIG. Administration of IVIG induced increased FoxP3 expression and inhibited Th17 cell development. The number of FoxP3+ Treg cells was increased, and the number of Th17 cells was decreased in the spleens of mice treated with IVIG. The number of FoxP3+ follicular helper T cells was increased, and subsequent maturation of germinal center B cells was inhibited by IVIG. In addition, IVIG up-regulated interleukin-10 (IL-10) and Fcγ receptor IIB expression. The treatment effects of IVIG on arthritis were lost in IL-10-knockout mice. These results showed that IVIG has therapeutic effects by modulating CD4+ T cell differentiation. The therapeutic effects of IVIG are dependent on IL-10.
Sigma-Aldrich
IL-17 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Interleukin-17A human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Serine, European Pharmacopoeia (EP) Reference Standard