Merck
  • UCP4A protects against mitochondrial dysfunction and degeneration in pink1/parkin models of Parkinson's disease.
UCP4A protects against mitochondrial dysfunction and degeneration in pink1/parkin models of Parkinson's disease.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2014-08-26)
Kai Wu, Jia Liu, Na Zhuang, Tao Wang
ABSTRACT
Genetic mutations in parkin or pink1 are the most common causes of familial Parkinson's disease. PINK1 and Parkin are components of a mitochondrial quality control pathway that degrades dysfunctional mitochondria via autophagy. Using a candidate gene approach, we discovered that overexpression of uncoupling protein 4A (ucp4A) suppresses a range of pink1 mutant phenotypes, including male sterility, locomotor defects, and muscle degeneration that result from abnormal mitochondrial morphology and function. Furthermore, UCP4A overexpression in pink1 mutants rescued mitochondria-specific phenotypes associated with mitochondrial membrane potential, production of reactive oxygen species, resistance to oxidative stress, efficiency of the electron transport chain, and mitochondrial morphology. Consistent with its role in protecting mitochondria, UCP4A rescued mitochondrial phenotypes of parkin mutant flies, as well. Finally, the genetic deletion of ucp4A resulted in increased sensitivity to oxidative stress, a phenotype that was enhanced by the loss of PINK1. Taken together, these results indicate that UCP4A prevents mitochondrial dysfunction and that modulation of UCP activity protects cells in a situation relevant for human Parkinson's disease.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.