Merck
  • IL2RA genetic variants reduce IL-2-dependent responses and aggravate human cutaneous leishmaniasis.
IL2RA genetic variants reduce IL-2-dependent responses and aggravate human cutaneous leishmaniasis.
Journal of immunology (Baltimore, Md. : 1950) (2015-02-13)
Pablo R S Oliveira, Hélia Dessein, Audrey Romano, Sandrine Cabantous, Maria E F de Brito, Ferrucio Santoro, Maira G R Pitta, Valéria Pereira, Lain C Pontes-de-Carvalho, Virmondes Rodrigues, Sima Rafati, Laurent Argiro, Alain J Dessein
ABSTRACT
The outcome of Leishmania infections varies substantially, depending on the host and the parasite strain; infection may be asymptomatic or cause mild or severe skin ulcers (cutaneous leishmaniasis [CL]), limited or disseminated lesions, or lethal visceral disease. We previously reported an association between IL-2R mutations and susceptibility to visceral leishmaniasis in children infected with Leishmania donovani. In the present study, we evaluated the possible role of IL-2 signaling in human CL. We first showed that the transcripts of several genes of the IL-2 pathway were abundant in skin lesions caused by Leishmania braziliensis. We then carried out a genetic analysis, focusing on major genes of the IL-2 pathway. We used a family-based approach and found that polymorphisms of several genes appeared to be associated with CL in a Brazilian population. Moreover, two polymorphisms of the IL2RA gene were significantly and independently associated with CL. We confirmed this result in a second Brazilian sample (also exposed to L. braziliensis) and in Iranians infected with Leishmania tropica: IL2RA rs10905669 T (Pcombined = 6 × 10(-7)) and IL2RA rs706778 T (Pcombined = 2 × 10(-9)) were associated with greater susceptibility to lesion development. These alleles were also correlated with a poor IFN-γ response and poor FOXP3(+) regulatory T cell activation. Thus, IL-2 plays a crucial role in protection against the cutaneous ulcers caused by Leishmania, and the IL-2 pathway is a potential target for strategies aiming to control Leishmania-related diseases.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology