Merck
  • Epigallocatechin-3-gallate suppresses cell proliferation and promotes apoptosis and autophagy in oral cancer SSC-4 cells.
Epigallocatechin-3-gallate suppresses cell proliferation and promotes apoptosis and autophagy in oral cancer SSC-4 cells.
OncoTargets and therapy (2015-03-12)
Alexandra Iulia Irimie, Cornelia Braicu, Oana Zanoaga, Valentina Pileczki, Claudia Gherman, Ioana Berindan-Neagoe, Radu Septimiu Campian
ABSTRACT
Epigallocatechin-3-gallate (EGCG) is the major bioactive component of green tea. Our experimental data indicated that EGCG treatment suppresses cell proliferation of SSC-4 human oral squamous cell carcinoma (OSCC), the effect being dose- and time-dependent. In parallel was observed the activation of apoptosis and autophagy, in response to EGCG exposure in SSC-4 cells. Treatment with EGCG activates the expression of the BAD, BAK, FAS, IGF1R, WNT11, and ZEB1 genes and inhibits CASP8, MYC, and TP53. All of these results suggest that EGCG has an excellent potential to become a therapeutic compound for patients with OSCC, by inducing tumor cell death via apoptosis and autophagy.
Sigma-Aldrich
Sodium succinate dibasic, purum, anhydrous, ≥98.0% (NT)
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Disodium succinate, ≥98%
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)