Merck
  • Liquid-phase exfoliated graphene as highly-sensitive sensor for simultaneous determination of endocrine disruptors: diethylstilbestrol and estradiol.
Liquid-phase exfoliated graphene as highly-sensitive sensor for simultaneous determination of endocrine disruptors: diethylstilbestrol and estradiol.
Journal of hazardous materials (2014-09-30)
Lintong Hu, Qin Cheng, Danchao Chen, Ming Ma, Kangbing Wu
ABSTRACT
It is quite important to develop convenient and rapid analytical methods for trace levels of endocrine disruptors because they heavily affect health and reproduction of humans and animals. Herein, graphene was easily prepared via one-step exfoliation using N-methyl-2-pyrrolidone as solvent, and then used to construct an electrochemical sensor for highly-sensitive detection of diethylstilbestrol (DES) and estradiol (E2). On the surface of prepared graphene film, two independent and greatly-increased oxidation waves were observed at 0.28V and 0.49V for DES and E2. The remarkable signal enlargements indicated that the detection sensitivity was improved significantly. The influences of pH value, amount of graphene and accumulation time on the oxidation signals of DES and E2 were discussed. As a result, a highly-sensitive and rapid electrochemical method was newly developed for simultaneous detection of DES and E2. The values of detection limit were evaluated to be 10.87 nM and 4.9 nM for DES and E2. Additionally, this new method was successfully used in lake water samples and the accuracy was satisfactory.
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Supelco
1-Methyl-2-pyrrolidinone, analytical standard
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%