Merck
  • Production of an anthocyanin-rich food colourant from Thymus moroderi and its application in foods.
Production of an anthocyanin-rich food colourant from Thymus moroderi and its application in foods.
Journal of the science of food and agriculture (2014-07-22)
Miriam Cristina Díaz-García, María Rosario Castellar, José María Obón, Concepción Obón, Francisco Alcaraz, Diego Rivera
ABSTRACT
Anthocyanins and other polyphenols from flowers and bracts of Thymus sp. are studied. An anthocyanin-rich food colourant with interesting high antioxidant activity from Thymus moroderi has been obtained, and applied to colour foods. Anthocyanins and other polyphenols from T. moroderi and another five Thymus sp. were extracted in methanol/hydrochloric acid 0.1 mol L(-1) (50/50, v/v) 2 h stirring at 50 °C. They were identified and quantified by HPLC-PDA-MS and UHPLC-PDA-fluorescence, as total individual polyphenols. Total polyphenols were also determined. Flowers had higher anthocyanins and other polyphenols concentrations than bracts; for example, total polyphenols content of T. moroderi were 131.58 and 61.98 g GAE kg(-1) vegetal tissue, respectively. A liquid concentrated colourant was obtained from T. moroderi using water/citric acid as solvent. It was characterised and compared with other two commercial anthocyanin-rich food colourants from red grape skin and red carrot (colour strength of 1.7 and 3.6 AU, respectively). T. moroderi colourant had 1.2 AU colour strength, and high storage stability (>97.1% remaining colour after 110 days at 4 °C). It showed a higher polyphenols content than commercial colourants. Its antioxidant activity was 0.707 mmol Trolox eq. g(-1) plant dry weight, 69.5 times higher than red carrot. The three colourants were applied to colour yogurts, giving pinky tonalities. The colour did not change evidently (ΔE*(ab)  < 3) when stored under refrigeration during 1 month. T. moroderi can be a source of anthocyanin-rich food colourant (E-163) with both high polyphenols content and high antioxidant activity. This colourant gives a stable colour to a yogurt during 1 month. These results expand the use of natural colourants.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Supelco
3,4-Dihydroxybenzoic acid, analytical standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid, 98%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
trans-Ferulic acid, ≥99%