Merck
  • Aflatoxins contamination in Pakistani brown rice: a comparison of TLC, HPLC, LC-MS/MS and ELISA techniques.
Aflatoxins contamination in Pakistani brown rice: a comparison of TLC, HPLC, LC-MS/MS and ELISA techniques.
Toxicology mechanisms and methods (2014-07-26)
Javed Iqbal, Muhammad Asif Asghar, Aftab Ahmed, Mobeen Ahmed Khan, Khalid Jamil
ABSTRACT
Advancement in the field of analytical food-chemistry has explored various experimental techniques for aflatoxins (AFs) quantification. The present study was aimed to compare four different techniques; thin-layer chromatography (TLC), high-performance liquid chromatography (HPLC), liquid chromatography-tandem mass spectrometry (LC-MS/MS) and enzyme-linked immunosorbent assay (ELISA) for the analysis of aflatoxin B₁ (AFB₁), B₂ (AFB₂), G₁ (AFG₁) and G₂ (AFG₂) in brown rice (n = 120) being collected from Karachi, Pakistan. All the four assays provide precised, accurate and comparable results. However, some differences were observed. For instance, TLC, HPLC and LC-MS/MS methodologies offered the advantage of the quantification of individual toxins in contrast to ELISA technique. The contamination ranges of AFB₁/AFB₂ as determined by TLC, HPLC and LC-MS/MS were 1.18-9.97/0.59-1.52, 0.16-10.54/0.26-1.35 and 0.11-10.88/0.38-1.48 µg/kg, respectively. However, AFG₁ and AFG₂ were not detected in any tested samples. Furthermore, owing to low-detection limit and sensitivity, HPLC and LC-MS/MS methodologies have identified greater number of contaminated samples in comparison to TLC and ELISA techniques. The overall average results of total AFs as provided by HPLC (3.79 µg/kg) and LC-MS/MS (3.89 µg/kg) were found higher in comparison to TLC (3.68 µg/kg) and ELISA (3.70 µg/kg). On the basis of achieved results, it was concluded that TLC, HPLC, LC-MS/MS and ELISA techniques are valuable tool for the quantification of AFs in cereals and grains. Furthermore, HPLC and LC-MS/MS techniques offer an added advantage for the detection of AFs in diminutive levels.
Product Description
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%