Merck
  • Research Resource: STR DNA profile and gene expression comparisons of human BG-1 cells and a BG-1/MCF-7 clonal variant.
Research Resource: STR DNA profile and gene expression comparisons of human BG-1 cells and a BG-1/MCF-7 clonal variant.
Molecular endocrinology (Baltimore, Md.) (2014-10-17)
Yin Li, Yukitomo Arao, Julie M Hall, Sandra Burkett, Liwen Liu, Kevin Gerrish, Vincent Cavailles, Kenneth S Korach
ABSTRACT
Human ovarian cancer BG-1 cells are a valuable in vitro model that has enabled several laboratories to study the estrogenic responses of ovarian cancers. We recently discovered that there are two different BG-1 cell lines being used for experiments, denoted here as BG-1 FR and BG-1 NIEHS, which exhibit striking morphological differences. The objective of this study was to methodically analyze these two BG-1 variants and compare their characteristics. Short tandem repeat analysis revealed that the DNA profile of BG-1 FR cells was unique, yet the Short tandem repeat pattern of BG-1 NIEHS was identical with that of MCF-7 cells. From a cytogenetic analysis, it became apparent that the BG-1 FR line had the same profile as previously reported, whereas the BG-1 NIEHS and MCF-7 cells share a similar genetic display. A significant number of unique chromosomal translocations were observed between the BG-1 NIEHS and MCF-7 cells, suggesting that acquired genotypic differences resulted in the formation of two lines from a common origin. Although all cell types demonstrated a similar estrogen responsiveness in reporter gene assays, a microarray analysis revealed distinct estrogen-responsive gene expression patterns with surprisingly moderate to low overlap. We conclude that BG-1 FR is the original ovarian cancer cell line, whereas the BG-1 NIEHS is a variant from the MCF-7 cells. These findings provide much needed clarification of the identities and characteristics of key cell line models that are widely used to study estrogen action in female reproductive cancers.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland