Merck
  • CELSR2-PSRC1-SORT1 gene expression and association with coronary artery disease and plasma lipid levels in an Asian Indian cohort.
CELSR2-PSRC1-SORT1 gene expression and association with coronary artery disease and plasma lipid levels in an Asian Indian cohort.
Journal of cardiology (2014-03-29)
Prathima Arvind, Jiny Nair, Srikarthika Jambunathan, Vijay V Kakkar, Jayashree Shanker
ABSTRACT
Genetic regulation of plasma lipids has been shown to influence the risk of coronary artery disease (CAD). We analyzed the relationship between rs599839 and rs646776 single nucleotide polymorphisms (SNPs) present in the CELSR2-PSRC1-SORT1 gene cluster, candidate gene expression, and their association with CAD and circulating lipid levels in a representative cohort of Asian Indians selected from the Indian Atherosclerosis Research Study. SNPs rs599839 and rs646776 were genotyped by Taqman assay in 1034 CAD patients (cases) and 1034 age- and gender-matched controls. Expression of CELSR2, PSRC1, and SORT1 genes was measured in 100 cases and 100 controls. Plasma levels of total cholesterol (TC), triglycerides, high-density lipoprotein-cholesterol, and low-density lipoprotein-cholesterol (LDL-c) were measured by enzymatic assay. Both rs646776 and rs599839 were in strong linkage disequilibrium (r = 0.98) and showed significant protective association with CAD (OR = 0.315, 95% CI 0.136-0.728, p<0.007 and OR = 0.422, 95% CI 0.181-0.981, p = 0.045, respectively). Haplotype TA showed 72% frequency and was associated with CAD (OR 0.77, 95% CI 0.67-0.88, p = 0.0002). PSRC1 gene expression was lower in the cases than in the controls (0.75 ± 0.405 versus 1.04 ± 0.622, p = 2.26 × 10(-4)). The homozygous variant and heterozygous genotypes showed 30% and 15% higher PSRC1 expression, respectively. Correspondingly, the minor alleles were associated with lower plasma TC and LDL-c levels. PSRC1 in the cholesterol gene cluster shows a significant association with CAD by virtue of the two SNPs, rs646776 and rs599839 that also regulate plasma cholesterol levels.
Product Description
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis