Merck
  • A grape seed extract increases active glucagon-like peptide-1 levels after an oral glucose load in rats.
A grape seed extract increases active glucagon-like peptide-1 levels after an oral glucose load in rats.
Food & function (2014-08-05)
Noemi González-Abuín, Neus Martínez-Micaelo, Maria Margalef, Mayte Blay, Anna Arola-Arnal, Begoña Muguerza, Anna Ardévol, Montserrat Pinent
ABSTRACT
We have previously reported that procyanidins, a class of flavonoids, improve glycemia and exert an incretin-like effect, which was linked to their proven inhibitory effect on the dipeptidyl-peptidase 4 (DPP4) activity. However, their actual effect on incretin levels has not been reported yet. Therefore, in the present study we have evaluated whether a grape seed extract enriched in procyanidins (GSPE) modulates plasma incretin levels and attempted to determine the mechanisms involved. An acute GSPE treatment in healthy Wistar female rats prior to an oral glucose load induced an increase in plasma active glucagon-like peptide-1 (GLP-1), which was accompanied by an increase in the plasma insulin/glucose ratio and a simultaneous decrease in glucose levels. In agreement with our previous studies, the intestinal DPP4 activity was inhibited by the GSPE treatment. We have also assayed in vitro whether this inhibition occurs in inner intestinal tissues close to GLP-1-producing cells, such as the endothelium of the capillaries. We have found that the main compounds absorbed by intestinal CaCo-2 cells after an acute treatment with GSPE are catechin, epicatechin, B2 dimer and gallic acid, and that they inhibit the DPP4 activity in endothelial HUVEC cells in an additive way. Moreover, an increase in plasma total GLP-1 levels was found, suggesting an increase in GLP-1 secretion. In conclusion, our results show that GSPE improves glycemia through its action on GLP-1 secretion and on the inhibition of the inner intestinal DPP4 activity, leading to an increase in active GLP-1 levels, which, in turn, may affect the insulin release.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Supelco
Procyanidin B2, analytical standard
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Methanol, analytical standard
Supelco
Acetone, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%